missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other OmpA S. Enteritidis Protein

A cDNA sequence encoding the OmpA S. Enteritidis was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12927-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15931979

  • 1740.00 € / 1mg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

OmpA S. Enteritidis Protein
Research Use Only
Recombinant Protein
E. coli
PBS.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
Protein is >95% pure as determined by 10% PAGE (coomassie staining).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers