missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Pfu DNA Polymerase Protein
A cDNA sequence encoding the Pfu DNA Polymerase was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13023-500U
Additional Details : Weight : 0.01000kg
Specifications
Pfu DNA Polymerase Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MILDVDYITE EGKPVIRLFK KENGKFKIEH DRTFRPYIYA LLRDDSKIEE VKKITGERHG KIVRIVDVEK VEKKFLGKPI TVWKLYLEHP QDVPTIREKV REHPAVVDIF EYDIPFAKRY LIDKGLIPME GEEELKILAF DIETLYHEGE EFGKGPIIMI SYADENEAKV ITWKNIDLPY VEVVSSEREM IKRFLRIIRE KDPDIIVTYN GDSFDFPYLA KRAEKLGIKL TIGRDGSEPK MQRIGDMTAV EVKGRIHFDL YHVITRTINL PTYTLEAVYE AIFGKPKEKV YADEIAKAWE SGENLERVAK YSMEDAKATY ELGKEFLPME IQLSRLVGQP LWDVSRSSTG NLVEWFLLRK AYERNEVAPN KPSEEEYQRR LRESYTGGFV KEPEKGLWEN IVYLDFRALY PSIIITHNVS PDTLNLEGCK NYDIAPQVGH KFCKDIPGFI PSLLGHLLEE RQKIKTKMKE TQDPIEKILL DYRQKAIKLL ANSFYGYYGY AKARWYCKEC AESVTAWGRK YIELVWKELE EKFGFKVLYI DTDGLYATIP GGESEEIKKK ALEFVKYINS KLPGLLELEY EGFYKRGFFV TKKRYAVIDE EGKVITRGLE IVRRDWSEIA KETQARVLET ILKHGDVEEA VRIVKEVIQK LANYEIPPEK LAIYEQITRP LHEYKAIGPH VAVAKKLAAK GVKIKPGMVI GYIVLRGDGP ISNRAILAEE YDPKKHKYDA EYYIENQVLP AVLRILEGFG YRKEDLRYQK TRQVGLTSWL NIKKS |
500 U | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
s-HCl (pH 8.8 at 25C), 100mM (NH4)2SO4, 100mM KCl, 1% Triton X-100, 1 mg/ml BSA, 20mM MgSO4. |