missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Pfu DNA Polymerase Protein

A cDNA sequence encoding the Pfu DNA Polymerase was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13023-2500U

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15922279

  • 1355.00 € / 2500 units
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Pfu DNA Polymerase Protein
Research Use Only
Recombinant Protein
E. coli
MILDVDYITE EGKPVIRLFK KENGKFKIEH DRTFRPYIYA LLRDDSKIEE VKKITGERHG KIVRIVDVEK VEKKFLGKPI TVWKLYLEHP QDVPTIREKV REHPAVVDIF EYDIPFAKRY LIDKGLIPME GEEELKILAF DIETLYHEGE EFGKGPIIMI SYADENEAKV ITWKNIDLPY VEVVSSEREM IKRFLRIIRE KDPDIIVTYN GDSFDFPYLA KRAEKLGIKL TIGRDGSEPK MQRIGDMTAV EVKGRIHFDL YHVITRTINL PTYTLEAVYE AIFGKPKEKV YADEIAKAWE SGENLERVAK YSMEDAKATY ELGKEFLPME IQLSRLVGQP LWDVSRSSTG NLVEWFLLRK AYERNEVAPN KPSEEEYQRR LRESYTGGFV KEPEKGLWEN IVYLDFRALY PSIIITHNVS PDTLNLEGCK NYDIAPQVGH KFCKDIPGFI PSLLGHLLEE RQKIKTKMKE TQDPIEKILL DYRQKAIKLL ANSFYGYYGY AKARWYCKEC AESVTAWGRK YIELVWKELE EKFGFKVLYI DTDGLYATIP GGESEEIKKK ALEFVKYINS KLPGLLELEY EGFYKRGFFV TKKRYAVIDE EGKVITRGLE IVRRDWSEIA KETQARVLET ILKHGDVEEA VRIVKEVIQK LANYEIPPEK LAIYEQITRP LHEYKAIGPH VAVAKKLAAK GVKIKPGMVI GYIVLRGDGP ISNRAILAEE YDPKKHKYDA EYYIENQVLP AVLRILEGFG YRKEDLRYQK TRQVGLTSWL NIKKS
2500 U
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
s-HCl (pH 8.8 at 25C), 100mM (NH4)2SO4, 100mM KCl, 1% Triton X-100, 1 mg/ml BSA, 20mM MgSO4.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers