missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other scIC2 Protein

A cDNA sequence encoding the scIC2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13433-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15933549

  • 203.00 € / 10µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

scIC2 Protein
Research Use Only
Recombinant Protein
E. coli
The protein solution contains 20mM Tris-HCl, 250mM NaCl and 50% glycerol, pH 7.5.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
Greater than 91.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers