missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other SODA Protein
A cDNA sequence encoding the SODA was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Name | SODA Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | Specific activity is > 350 units/mg, in which one unit will inhibit the rate of reduction of cytochrome c by 50% in a coupled system, using xanthine and xanthine oxidase at pH 7.8 at 25C in a 1.5 ml reaction volume. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15975408
|
enQuireBio™
QP10867-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965408
|
enQuireBio™
QP10867-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955408
|
enQuireBio™
QP10867-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
SODA Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSYTLPSLPY AYDALEPHFD KQTMEIHHTK HHQTYVNNAN AALESLPEFA NLPVEELITK LDQLPADKKT VLRNNAGGHA NHSLFWKGLK KGTTLQGDLK AAIERDFGSV DNFKAEFEKA AASRFGSGWA WLVLKGDKLA VVSTANQDSP LMGEAISGAS GFPIMGLDVW EHAYYLKFQN RRPDYIKEFW NVVNWDEAAA RFAAKK | |
Greater than 95% as determined by SDS-PAGE. |
Research Use Only | |
Specific activity is > 350 units/mg, in which one unit will inhibit the rate of reduction of cytochrome c by 50% in a coupled system, using xanthine and xanthine oxidase at pH 7.8 at 25C in a 1.5 ml reaction volume. | |
Other | |
His | |
SODA E. coli solution (1 mg/ml) containining 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol and 0.1M NaCl. |