missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Streptavidin Protein

A cDNA sequence encoding the Streptavidin was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13625-20mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15904189

  • 203.00 € / 20mg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Streptavidin Protein
Research Use Only
Recombinant Protein
Non-species Specific
MRKIVVAAIA VSLTTVSITA SASADPSKDS KAQVSAAEAG ITGTWYNQLG STFIVTAGAD GALTGTYESA VGNAESRYVL TGRYDSAPAT DGSGTALGWT VAWKNNYRNA HSATTWSGQY VGGAEARINT QWLLTSGTTE ANAWKSTLVG HDTFTKVKPS AASIDAAKKA GVNNGNPLDA VQQ
20 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
Lyophilized (1 mg/ml) in 50mM NaCl, pH 9.0.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers