missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other T. pallidum p41 Protein
A cDNA sequence encoding the T. pallidum p41 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13673-1mg
Additional Details : Weight : 0.01000kg
Specifications
T. pallidum p41 Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged |
Treponema pallidum protein is >90% pure as determined by 10% PAGE (coomassie staining). | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
10mM Tris-HCl pH 8.0, 1mM EDTA, 1mMDTT and 8M urea. |