missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other T4 DNA Protein
A cDNA sequence encoding the T4 DNA was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10872-500,000IU
Additional Details : Weight : 0.01000kg
Specifications
T4 DNA Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
50mM KCl, 10mM Tris-HCl (pH 7.4), 0.1mM EDTA, 1mM DTT, 200 microg/ml BSA and 50% glycerol. Store at -20C. |
500,000 IU | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Other | |
Untagged |