missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Terlipressin Protein

A cDNA sequence encoding the Terlipressin was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13723-250mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15944499

  • 1015.00 € / 250mg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Terlipressin Protein
Research Use Only
Recombinant Protein
E. coli
The protein (1 mg/ml) was lyophilized with no additives.
250 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers