missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Urease Protein
Click to view available options
Quantity:
1 mg
5 mg
50 mg
Unit Size:
1mg
50mg
5mg
Specifications
Specifications
| Name | Urease Protein |
| Quantity | 1 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
| Biological Activity | The activity was found to be 141U/mg powder. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Other |
| Species | E. coli |
| Protein Tag | Untagged |
| Buffer | Each mg of protein contains 420microg Potassium Phosphate and 30microg EDTA Na2. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction