missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Ovine GH Protein
A cDNA sequence encoding the GH was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10634-100ug
Additional Details : Weight : 0.01000kg
Specifications
GH Protein | |
Research Use Only | |
Ovine placental Grwoth Hormone is fully biologically active when compared to World Health Organization (WHO) reference standard using in vitro bioassay in PDF-P1 3B9 cells stably transfected with rabbit GH receptors. It is also capable of forming a 1:2 complex with the recombinant ovine growth hormone receptor extracellular domain (ECD). | |
Ovine | |
Untagged | |
Greater than 98.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE. |
100 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.0045mM NaHCO3 adjusted to pH 9. |