missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Ovine Prolactin Protein
A cDNA sequence encoding the Prolactin was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10851-EC-10ug
Additional Details : Weight : 0.01000kg
Specifications
Prolactin Protein | |
Research Use Only | |
Is fully biologically active as evidenced by inducing proliferation of Nb2 cells. | |
Ovine | |
Untagged | |
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.0045mM NaHCO3. |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MDSKGSAQKG SRLLLLLVVS NLLLCQGVVS TPVCPNGPGN CQVSLRDLFD RAVMVSHYIH NLSSEMFNEF DKRYAQGKGF ITMALNSCHT SSLPTPEDKE QAQQTHHEVL MSLILGLLRS WNDPLYHLVT EVRGMKGVPD AILSRAIEIE EENKRLLEGM EMIFGQVIPG AKETEPYPVW SGLPSLQTKD EDARHSAFYN LLHCLRRDSS KIDTYLKLLN CRIIYNNNC | |
Greater than 98.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE. |