Learn More
enQuireBio™ Recombinant P. aeruginosa Protease LasA Protein
A DNA sequence encoding the Pseudomonas aeruginosa (strain ATCC 15692 / PAO1 / 1C / PRS 101 / LMG 12228) Protease LasA, was expressed in the hosts and tags indicated. option, ab
Brand: enQuireBio™ QP7030-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_250562.1 | |
P. aeruginosa Protease LasA Protein | |
500 μg | |
Not determined. | |
Full Length of Mature Protein | |
P. aeruginosa | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
878260 | |
Greater than 80% as determined by SDS-PAGE. | |
Research Use Only | |
lasA | |
Recombinant Protein | |
E. coli | |
APPSNLMQLP WRQGYSWQPN GAHSNTGSGY PYSSFDASYD WPRWGSATYS VVAAHAGTVR VLSRCQVRVT HPSGWATNYY HMDQIQVSNG QQVSADTKLG VYAGNINTAL CEGGSSTGPH LHFSLLYNGA FVSLQGASFG PYRINVGTSN YDNDCRRYYF YNQSAGTTHC AFRPLYNPGL AL |