missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant P. aeruginosa Protease LasA Protein
A DNA sequence encoding the Pseudomonas aeruginosa (strain ATCC 15692 / PAO1 / 1C / PRS 101 / LMG 12228) Protease LasA, was expressed in the hosts and tags indicated. option, ab
436.00 € - 3590.00 €
Specifications
Accession Number | NP_250562.1 |
---|---|
Gene ID (Entrez) | 878260 |
Name | P. aeruginosa Protease LasA Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16007110
|
enQuireBio™
QP7030-100UG |
100 μg |
1025.00 €
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16027110
|
enQuireBio™
QP7030-1MG |
1 mg |
3590.00 €
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16017110
|
enQuireBio™
QP7030-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16037110
|
enQuireBio™
QP7030-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16047110
|
enQuireBio™
QP7030-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16057110
|
enQuireBio™
QP7030-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_250562.1 | |
P. aeruginosa Protease LasA Protein | |
Not determined. | |
Full Length of Mature Protein | |
P. aeruginosa | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
878260 | |
Research Use Only | |
lasA | |
Recombinant Protein | |
E. coli | |
APPSNLMQLP WRQGYSWQPN GAHSNTGSGY PYSSFDASYD WPRWGSATYS VVAAHAGTVR VLSRCQVRVT HPSGWATNYY HMDQIQVSNG QQVSADTKLG VYAGNINTAL CEGGSSTGPH LHFSLLYNGA FVSLQGASFG PYRINVGTSN YDNDCRRYYF YNQSAGTTHC AFRPLYNPGL AL | |
Greater than 80% as determined by SDS-PAGE. |