missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant P. heparinus Chondroitinase AC Protein

Product Code. 18799267
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18799267 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18799267 Supplier R&D Systems™ Supplier No. 8384GH050
  • ONLINE EXCLUSIVE PRICE
    509.50 €
    453.60 € / 50µg
    Save 55.90 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to hydrolyze chondroitin sulfate. The specific activity is >50,000 pmol/min/μg, as measured under the described conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q59288
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl and Brij-35.
Gene ID (Entrez) 8251875
Molecular Weight (g/mol) M.W. (Observed): 66 kDa, reducing conditions; M.W. (theoretical): 78 kDa
Name Chondroitinase AC
Quality Control Testing SDS-PAGE: 66kDa, reducing conditions
Quantity 50 μg
Source E. coli-derived p. heparinus Chondroitinase AC protein Gln23-Lys700 with an N-terminal Met and 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Regulatory Status RUO
Termination Met
Without Additives Carrier Protein
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Symbol Phep_0786
Conjugate Unconjugated
Recombinant Recombinant
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain (at 5 μg / lane)
Protein Chondroitinase AC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.