missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Pig C5a / Complement 5a Protein

A DNA sequence encoding the Sus scrofa (Pig) C5a / Complement 5a, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9020-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16037471

  • 1050.00 € / 100µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

C5a / Complement 5a Protein
Research Use Only
C5
Recombinant Protein
Yeast
MLQKKIEEEA AKYKYAMLKK CCYDGAYRND DETCEERAAR IKIGPKCVKA FKDCCYIANQ VRAEQSHKNI QLGR
Greater than 90% as determined by SDS-PAGE.
100 μg
Not determined.
Full Length
Pig
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers