missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pig HSP90AA1 / HSP90 Protein
A DNA sequence encoding the Sus scrofa (Pig) HSP90AA1 / HSP90, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP10039-ba-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_999138.1 | |
HSP90AA1 / HSP90 Protein | |
Research Use Only | |
HSP90AA1 | |
Recombinant Protein | |
Baculovirus | |
VEKERDKEVS DDEAEEKEDK EEEKEKEEKE SEDKPEIEDV GSDEEEEEKK DGDKKKKKKI KEKYIDQEEL NKTKPIWTRN PDDITNEEYG EFYKSLTNDW EDHLAVKHFS VEGQLEFRAL LFVPRRAPFD LFENRKKKNN IKLYVR | |
Greater than 90% as determined by SDS-PAGE. |
397028 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Pig | |
His | |
Tris-based buffer, 50% glycerol |