missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Porcine FSH Protein
A cDNA sequence encoding the FSH was constructed and used to recombinantly synthesize the protein.
131.00 € - 570.00 €
Specifications
Name | FSH Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Porcine |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15958748
|
enQuireBio™
QP11904-100IU |
100 IU |
131.00 €
100 units |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15968748
|
enQuireBio™
QP11904-5000IU |
5000 IU |
570.00 €
5000 units |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15978748
|
enQuireBio™
QP11904-500IU |
500 IU |
336.00 €
500 units |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
FSH Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Porcine | |
Untagged | |
The FSH was lyophilized with no additives. |
Research Use Only | |
Recombinant Protein | |
Porcine | |
MKSLQFCFLF CCWKAICCNS CELTNITITV EKEECNFCIS INTTWCAGYC YTRDLVYKDP ARPNIQKTCT FKELVYETVK VPGCAHHADS LYTYPVATEC HCGKCDSDST DCTVRGLGPS YCSFSEMKE |