missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Porcine Insulin Protein

Product Code. 15915008
Click to view available options
Quantity:
25 mg
250 mg
1 g
Unit Size:
1g
250mg
25mg
This item is not returnable. View return policy

Product Code. 15915008

Brand: enQuireBio™ QP107331gr

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the Insulin was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 397415
Name Insulin Protein
Quantity 1 g
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Gene Symbol INS
Biological Activity >27 units/mg.
Product Type Recombinant Protein
Cross Reactivity Porcine
Species Porcine
Protein Tag Untagged
Sequence MALWTRLLPL LALLALWAPA PAQAFVNQHL CGSHLVEALY LVCGERGFFY TPKARREAEN PQAGAVELGG GLGGLQALAL EGPPQKRGIV EQCCTSICSL YQLENYCN
Buffer The Insulin Porcine was lyophilized from a concentrated (1 mg/ml) solution with no additives.
Purity or Quality Grade Greater than 97.0% as determined by SEC-HPLC.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.