missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Porcine PD-L1/B7-H1 Fc Chimera Protein
Click to view available options
Quantity:
100 μg
Unit Size:
50µg
Description
Measured by its binding ability in a functional ELISA. When Recombinant Porcine PD-L1/B7-H1 Fc Chimera (Catalog # 10397-B7) is immobilized at 2 μg/mL (100 μL/well), the concentration of Recombinant Porcine PD-1 His-tag that produces 50% of the optimal binding response is found to be approximately 0.8-6 μg/mL.
Specifications
Specifications
| Accession Number | NP_001020392 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 29126 |
| Molecular Weight (g/mol) | M.W. (Observed): 65-85 kDa, under reducing conditions; M.W. (theoretical): 52 kDa |
| Quantity | 100 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived porcine PD-L1/B7-H1 protein Porcine PD-L1/B7-H1 (Phe19-Thr237) (N-terminus) Accession # NP_001020392.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Show More |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido