missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rabbit Tissue factor pathway inhibitor Protein
A DNA sequence encoding the Oryctolagus cuniculus (Rabbit) Tissue factor pathway inhibitor, was expressed in the hosts and tags indicated.
352.00 € - 3175.00 €
Specifications
Accession Number | NP_001095184.1 |
---|---|
Gene ID (Entrez) | 100009401 |
Name | Tissue factor pathway inhibitor Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16095750
|
enQuireBio™
QP6775-YE-100UG |
100 μg |
1050.00 €
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16035760
|
enQuireBio™
QP6775-YE-500UG |
500 μg |
2535.00 €
500µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16005760
|
enQuireBio™
QP6775-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16025760
|
enQuireBio™
QP6775-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16045760
|
enQuireBio™
QP6775-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16015760
|
enQuireBio™
QP6775-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001095184.1 | |
Tissue factor pathway inhibitor Protein | |
Not determined. | |
Full Length | |
Rabbit | |
His | |
Tris-based buffer, 50% glycerol |
100009401 | |
Research Use Only | |
TFPI | |
Recombinant Protein | |
Yeast | |
AAEEDEEFTN ITDIKPPLQK PTHSFCAMKV DDGPCRAYIK RFFFNILTHQ CEEFIYGGCE GNENRFESLE ECKEKCARDY PKMTTKLTFQ KGKPDFCFLE EDPGICRGYI TRYFYNNQSK QCERFKYGGC LGNLNNFESL EECKNTCENP TSDFQVDDHR TQLNTVNNTL INQPTKAPRR WAFHGPSWCL PPADRGLCQA NEIRFFYNAI IGKCRPFKYS GCGGNENNFT SKKACITACK KGFIPKSIKG GLIKTKRKKK KQPVKITYVE TFVKKT | |
Greater than 90% as determined by SDS-PAGE. |