missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat A2M / CPAMD5 / Alpha-2-macroglobulin Protein
A DNA sequence encoding the Rattus norvegicus (Rat) A2M / CPAMD5 / Alpha-2-macroglobulin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8951-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_036620.2 | |
A2M / CPAMD5 / Alpha-2-macroglobulin Protein | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
100911545 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
A2m | |
Recombinant Protein | |
Yeast | |
LIYDLLPVKD LTGFPQGADQ REEDTNGCVK QNDTYINGIL YSPVQNTNEE DMYGFLKDMG LKVFTNSNIR KPKVCERLRD NKGIPAAYHL VSQSHMDAFL ESSESPTETR RSYFPETWIW DLVVVDSAGV A |