missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Activin A / INHBA Protein
Recombinant Protein
Brand: enQuireBio™ QP5498-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_058824.1 | |
Activin A / INHBA Protein | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 10 ng/mL in the Cytotoxicity of MPC-11 cells assay. The bioactivity acceptance criteria is set at 1 x 10^5 units/mg. | |
Rat | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
29200 | |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
INHBA | |
Recombinant Protein | |
E. coli | |
MGLECDGKVN ICCKKQFFVS FKDIGWNDWI IAPSGYHANY CEGECPSHIA GTSGSSLSFH STVINHYRMR GHSPFANLKS CCVPTKLRPM SMLYYDDGQN IIKKDIQNMI VEECGCS |