missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Adipsin / Complement Factor D / CFD Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Adipsin / Complement Factor D / CFD, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5823-ma-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15948019

  • 1850.00 € / 100µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001071110.1
Adipsin / Complement Factor D / CFD Protein
Research Use Only
Cfd
Recombinant Protein
Mammal
ILGGQEAMAH ARPYMASVQV NGTHVCGGTL VDEQWVLSAA HCMDGVTKDE VVQVLLGAHS LSSPEPYKHL YDVQSVVLHP GSRPDSVEDD LMLFKLSHNA SLGPHVRPLP LQREDREVKP GTLCDVAGWG VVTHAGRRPD VLQQLTVSIM DRNTCNLRTY HDGAITKNMM CAESNRRDTC RGDSGGPLVC GDAVEAVVTW GSRVCGNRRK PGVFTRVATY VPWIENVLSG NVSVNVTA
Greater than 82.10% as determined by SDS-PAGE.
54249
100 μg
Not determined.
Full Length of Mature Protein
Rat
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers