missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Cyclophilin F Protein
A cDNA sequence encoding the Cyclophilin F was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11581-20ug
Additional Details : Weight : 0.01000kg
Specifications
282819 | |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Ppif | |
Rat | |
His | |
Cyclophilin F protein solution (1 mg/ml) containing Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT. |
Cyclophilin F Protein | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSCSDGGAR GANSSSQNPL VYLDVGADGQ PLGRVVLELK ADVVPKTAEN FRALCTGEKG FGYKGSTFHR VIPAFMCQAG DFTNHNGTGG KSIYGSRFPD ENFTLKHVGP GVLSMANAGP NTNGSQFFIC TIKTDWLDGK HVVFGHVKEG MDVVKKIESF GSKSGKTSKK IVITDCGQLS |