missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat DPP4 / CD26 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) DPP4 / CD26, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5944-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_036921.1 | |
DPP4 / CD26 Protein | |
Research Use Only | |
Dpp4 | |
Recombinant Protein | |
E. coli | |
SMVLGSGSGV FKCGIAVAPV SRWEYYDSVY TERYMGLPTP EDNLDHYRNS TVMSRAENFK QVEYLLIHGT ADDNVHFQQS AQISKALVDA GVDFQAMWYT DEDHGIASST AHQHIYSHMS HFLQQCFSLR | |
Greater than 90% as determined by SDS-PAGE. |
25253 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |