missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat FABP3 / H-FABP Protein
A DNA sequence encoding the Rattus norvegicus (Rat) FABP3 / H-FABP, was expressed in the hosts and tags indicated.
580.00 € - 1620.00 €
Specifications
Accession Number | NP_077076.1 |
---|---|
Gene ID (Entrez) | 79131 |
Name | FABP3 / H-FABP Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15929369
|
enQuireBio™
QP6013-MA-20UG |
20 μg |
580.00 €
20µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15919369
|
enQuireBio™
QP6013-MA-100UG |
100 μg |
1620.00 €
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15939369
|
enQuireBio™
QP6013-MA-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_077076.1 | |
FABP3 / H-FABP Protein | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
79131 | |
Research Use Only | |
Fabp3 | |
Recombinant Protein | |
Mammal | |
ADAFVGTWKL VDSKNFDDYM KSLGVGFATR QVASMTKPTT IIEKNGDTIT IKTHSTFKNT EISFQLGVEF DEVTADDRKV KSVVTLDGGK LVHVQKWDGQ ETTLTRELSD GKLILTLTHG NVVSTRTYEK EA | |
Greater than 90% as determined by SDS-PAGE. |