missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Fetuin-B / FETUB Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Fetuin-B / FETUB, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9897-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_445800.2 | |
Fetuin-B / FETUB Protein | |
Research Use Only | |
Fetub | |
Recombinant Protein | |
Yeast | |
RSPPAPPLPN APFAPLRPLG CNDSEVLAVA GFALQNINRV QKDGYMLTLN RVHDARVHRQ EDMGSLFYLM LDVLETGCHV LSRKALKDCG PRIFYETVHG QCKAMFHVNK PRRVLYLPAY NCTLRPVSKR KIHSMCPDCP HPVDLSAPSV LEAATESLAK FNSENPSKQY ALVKVTKATT QWVVGPSYFV EYLIKESPCT QSQDSCSLQA SDSEPVGLCQ GSLIKSPGVP PQRFKKTVTV SCEFFESQDQ VPGGENPADT QDAKKLPQKN TAPTSSPSIT APRGSIQHLP EQEEPEDSKG KSPEEPFPVQ LDLTTNPQGD TLDVSFLYLE PEEKKLVVLP FPGKEQRSPE CPGPEKQRTP | |
Greater than 90% as determined by SDS-PAGE. |
83928 | |
100 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |