missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat FGF-18 Protein
A cDNA sequence encoding the FGF-18 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10602-5ug
Additional Details : Weight : 0.01000kg
Specifications
FGF-18 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
EENVDFRIHV ENQTRARDDV SRKQLRLYQL YSRTSGKHIQ VLGRRISARG EDGDKYAQLL VETDTFGSQV RIKGKETEFY LCMNRKGKLV GKPDGTSKEC VFIEKVLENN YTALMSAKYS GWYVGFTKKG RPRKGPKTRE NQQDVHFMKR YPKGQTELQK PFKYTTVTKR SRRIRPTHPG | |
Greater than 95.0% as determined by SDS-PAGE and HPLC analyses. |
5 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Rat | |
Untagged | |
Lyophilized from a 0.2um filtered concentrated solution in PBS, pH 7.4. |