missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Galectin-3-binding Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Galectin-3-binding protein, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Accession Number | NP_620796.1 |
---|---|
Gene ID (Entrez) | 245955 |
Name | Galectin-3-binding Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16022260
|
enQuireBio™
QP6296-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16042260
|
enQuireBio™
QP6296-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16032260
|
enQuireBio™
QP6296-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16052260
|
enQuireBio™
QP6296-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16062260
|
enQuireBio™
QP6296-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16072260
|
enQuireBio™
QP6296-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_620796.1 | |
Galectin-3-binding Protein | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
245955 | |
Research Use Only | |
Lgals3bp | |
Recombinant Protein | |
E. coli | |
AKDGDMRLVN GASASEGRVE IFYRGRWGTV CDNLWNLLDA HVVCRALGYE NATQALSRAA FGPGKGPIML DEVECTGNES SLANCSSLGW MVSHCGHEKD AGVVCSNDSR GIHILDLSGE LPDALGQIFD SQQDCDLFIQ VTGQGHGDLS LCAHTLILRT NPEAQALWQV VGSSVIMRVD AECMPVVRDF LRYFYSRRIE VSMSSVKCLH KLASAYGATE LQGYCGRLFV TLLPQDPTFH TPLELYEYAQ ATGDSVLEDL CVQFLAWNFE PLTQAEAWLS VPNALIQALL PKSELAVSSE LDLLKAVDQW STATGASHGD VERLVEQIRF PMMLPQELFE LQFNLSLYQG HQALFQRKTM EALEFHTVPL KVLAKYRSLN LTEDVYKPRL YTSSTWSSLL MAGAWSTQSY KYRQFYTYNY GSQSRYSSYQ NFQTPQHPSF LFKDKLISWS ATYLPTIQSC WNYGFSCTSD ELPVLGLTTS SYSDPTIGYE NKALILCGGY SVVDVTTFIG SKAPIPGTQE TNSSKTPSLF PCASGAFSSF RVVIRPFYLT NSTDTE | |
Greater than 90% as determined by SDS-PAGE. |