missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat GH Protein

Product Code. 15918948
Click to view available options
Quantity:
1 mg
100 μg
200 μg
Unit Size:
100µg
1mg
200µg
This item is not returnable. View return policy

Product Code. 15918948

Brand: enQuireBio™ QP11964100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the GH was constructed and used to recombinantly synthesize the protein.

Specifications

Name GH Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Rat
Species E. coli
Protein Tag Untagged
Sequence MAADSQTPWL LTFSLLCLLW PQEAGAFPAM PLSSLFANAV LRAQHLHQLA ADTYKEFERA YIPEGQRYSI QNAQAAFCFS ETIPAPTGKE EAQQRTDMEL LRFSLLLIQS WLGPVQFLSR IFTNSLMFGT SDRVYEKLKD LEEGIQALMQ ELEDGSPRIG QILKQTYDKF DANMRSDDAL LKNYGLLSCF KKDLHKAETY LRVMKCRRFA ESSCAF
Buffer The protein (1 mg/ml) was lyophilized after extensive dialyses against 5mM phosphate buffer, 5 mg mannitol and 1 mg glycine.
Purity or Quality Grade Greater than 97.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.