missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat GHBP Protein

Product Code. 15928958
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
This item is not returnable. View return policy

Product Code. 15928958

Brand: enQuireBio™ QP119672ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the GHBP was constructed and used to recombinantly synthesize the protein.

Specifications

Name GHBP Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Rat
Species Baculovirus
Protein Tag His
Sequence FPGSGATPAT LGKASPVLQR INPSLRESSS GKPRFTKCRS PELETFSCYW TEGDDHNLKV PGSIQLYYAR RIAHEWTPEW KECPDYVSAG ANSCYFNSSY TSIWIPYCIK LTTNGDLLKE KCFTVDEIVQ PDPPIGLNWT LLNISLPGIR GDIQVSWQPP PSADVLKGWI ILEYEIQYKE VNETKWKTMS PIWSTSVPLY SLRLDKEHEV RVRSRQRSFE KYSEFSEVLR VTFPQMDTLA ACEEDFRLEH HHHHH
Buffer GHBP protein solution (0.5 mg/ml) contains phosphate buffered saline (pH 7.4) and 10% glycerol.
Purity or Quality Grade Greater than 95.0% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.