Learn More
enQuireBio™ Recombinant Rat GM-CSF / CSF2 Protein
Recombinant Protein
Brand:  enQuireBio™ QP5495-1mg
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15946249
  • 4205.00 € / 1mg
Estimated Shipment: 21-06-2024
to see stock.
Specifications
NP_446304.1
GM-CSF / CSF2 Protein
Research Use Only
CSF2
Recombinant Protein
E. coli
MAPTRSPNPV TRPWKHVDAI KEALSLLNDM RALENEKNED VDIISNEFSI QRPTCVQTRL KLYKQGLRGN LTKLNGALTM IASHYQTNCP PTPETDCEIE VTTFEDFIKN LKGFLFDIPF DCWKPVQK
Greater than 95% as determined by SDS-PAGE.
116630
1 mg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Shows bioactivity at a concentration of 20 pg/mL in the FDCP-1 cell proliferation assay. The bioactivity acceptance criteria is set at 5.0 x 10^7 units/mg.
Rat
Untagged
20 mM sodium bicarbonate buffer, pH 8.5