missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat GM-CSF / CSF2 Protein
Recombinant Protein
Brand: enQuireBio™ QP5495-20ug
Additional Details : Weight : 0.01000kg
Specifications
NP_446304.1 | |
GM-CSF / CSF2 Protein | |
20 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 20 pg/mL in the FDCP-1 cell proliferation assay. The bioactivity acceptance criteria is set at 5.0 x 10^7 units/mg. | |
Rat | |
Untagged | |
20 mM sodium bicarbonate buffer, pH 8.5 |
116630 | |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
CSF2 | |
Recombinant Protein | |
E. coli | |
MAPTRSPNPV TRPWKHVDAI KEALSLLNDM RALENEKNED VDIISNEFSI QRPTCVQTRL KLYKQGLRGN LTKLNGALTM IASHYQTNCP PTPETDCEIE VTTFEDFIKN LKGFLFDIPF DCWKPVQK |