missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat IL 1 alpha Protein

A cDNA sequence encoding the IL 1 alpha was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12390-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15990259

  • 3915.00 € / 1mg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

24493
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSSAPHSFQ NNLRYKLIRI VKQEFIMNDS LNQNIYVDMD RIHLKAASLN DLQLEVKFDM YAYSSGGDDS KYPVTLKVSN TQLFVSAQGE DKPVLLKEIP ETPKLITGSE TDLIFFWEKI NSKNYFTSAA FPELLIATKE QSQVHLARGL PSMIDFQIS
Greater than 95.0% as determined by SDS-PAGE.
IL 1 alpha Protein
Research Use Only
Il1a
Rat
His
IL 1 alpha protein solution (1 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers