missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat IL-1 beta / IL1B Protein

Product Code. p-7152163
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
500 μg
Unit Size:
100µg
10µg
1mg
2µg
500µg
This item is not returnable. View return policy

Product Code. 15946319

Brand: enQuireBio™ QP55162ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Specifications

Name IL-1 beta / IL1B Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Gene Symbol IL1B
Biological Activity Rat IL-1 beta / IL1B induces D10S cell proliferation (typical ED50 is ? 25 ng/ml).
Product Type Recombinant Protein
Cross Reactivity Rat
Species E. coli
Protein Tag Untagged
Sequence MVPIRQLHCR LRDEQQKCLV LSDPCELKAL HLNGQNISQQ VVFSMSFVQG ETSNDKIPVA LGLKGLNLYL SCVMKDGTPT LQLESVDPKQ YPKKKMEKRF VFNKIEVKTK VEFESAQFPN WYISTSQAEH RPVFLGNSNG RDIVDFTMEP VSS
Buffer 10 mM sodium phosphate, pH 7.5
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.