missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL-17AF Protein
Recombinant Protein
107.00 € - 7105.00 €
Specifications
Name | IL-17AF Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Gene Symbol | IL17 |
Biological Activity | Rat IL-17AF induces IL-6 production in 3T3 cells as detected via ELISA (each lot is tested and typical ED50 is less than 20 ng/ml). |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15925679
|
enQuireBio™
QP5206-5UG |
5 μg |
107.00 €
5µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15905679
|
enQuireBio™
QP5206-25UG |
25 μg |
252.00 €
25µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15985669
|
enQuireBio™
QP5206-100UG |
100 μg |
940.00 €
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15915679
|
enQuireBio™
QP5206-500UG |
500 μg |
4325.00 €
500µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15995669
|
enQuireBio™
QP5206-1MG |
1 mg |
7105.00 €
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
IL-17AF Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Rat IL-17AF induces IL-6 production in 3T3 cells as detected via ELISA (each lot is tested and typical ED50 is less than 20 ng/ml). | |
Rat | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
Research Use Only | |
IL17 | |
Recombinant Protein | |
E. coli | |
IL-17A:MAV LIPQSSVCPN AEANNFLQNV KVNLKVLNSL SSKASSRRPS DYLNRSTSPW TLSRNEDPDR YPSVIWEAQC RHQRCVNAEG KLDHHMNSVL IQQEILVLKR EPEKCPFTFR VEKMLVGVGC TCVSSIVRHA S IL-17F:MA RRNPKVGLSA LQKAGNCPPL EDNSVRVDIR IFNQNQGISV PRDFQNRSSS PWDYNITRDP DRFPSEIAEA QCRHSGCINA QGQEDGSMNS VPIQQEILVL RREPQGCSNS FRLEKMLIKV GCTCVTPIVH HAA | |
Greater than 95% as determined by SDS-PAGE. |