missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL 4 Protein
A cDNA sequence encoding the IL 4 was constructed and used to recombinantly synthesize the protein.
131.00 € - 4205.00 €
Specifications
Gene ID (Entrez) | 287287 |
---|---|
Name | IL 4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | Il4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15994948
|
enQuireBio™
QP10716-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15984948
|
enQuireBio™
QP10716-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15974948
|
enQuireBio™
QP10716-1MG |
1 mg |
4205.00 €
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
287287 | |
Research Use Only | |
Il4 | |
Recombinant Protein | |
E. coli | |
HGCNDSPLR EIINTLNQVT EKGTPCTEMF VPDVLTATRN TTENELICRA SRVLRKFYFP RDVPPCLKNK SGVLGELRKL CRGVSGLNSL RSCTVNESTL TTLKDFLESL KSILRGKYLQ SCTSMS | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
IL 4 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 as determined in a cell proliferation assay using rat splenocytes is less than 2ng/ml corresponding to a specific activity of 500,000IU/mg. | |
Rat | |
Untagged | |
Lyophilized from a 0.2microm filtered concentrated solution in PBS pH 7.4 and 5% trehalose. |