missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Kcne2 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Kcne2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6254-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031950

  • 735.00 € / 100µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_598287.1
Kcne2 Protein
Research Use Only
Kcne2
Recombinant Protein
E. coli
MTTLANLTQT LEDAFKKVFI TYMDSWRRNT TAEQQALQAR VDAENFYYVI LYLMVMIGMF AFIVVAILVS TVKSKRREHS QDPYHQYIVE DWQQKYRSQI LHLEDSKATI HENLGATGFT VSP
Greater than 90% as determined by SDS-PAGE.
171138
100 μg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers