missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat MANF Protein
A cDNA sequence encoding the MANF was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10780-20ug
Additional Details : Weight : 0.01000kg
Specifications
MANF Protein | |
Research Use Only | |
The ED50 as determined by a cell proliferation assay using rat C6 cells is less than 10microg/ml, corresponding to a specific activity of >100 IU/mg. | |
Rat | |
Untagged | |
Lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |
20 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
LRPGDCEVCI SYLGRFYQDL KDRDVTFSPA TIEEELIKFC REARGKENRL CYYIGATDDA ATKIINEVSK PLAHHIPVEK ICEKLKKKDS QICELKYDKQ IDLSTVDLKK LRVKELKKIL DDWGEMCKGC AEKSDYIRKI NELMPKYAPK AASARTDL | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |