missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat MANF Protein

A cDNA sequence encoding the MANF was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10780-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15965148

  • 131.00 € / 5µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

MANF Protein
Research Use Only
The ED50 as determined by a cell proliferation assay using rat C6 cells is less than 10microg/ml, corresponding to a specific activity of >100 IU/mg.
Rat
Untagged
Lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4.
5 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
LRPGDCEVCI SYLGRFYQDL KDRDVTFSPA TIEEELIKFC REARGKENRL CYYIGATDDA ATKIINEVSK PLAHHIPVEK ICEKLKKKDS QICELKYDKQ IDLSTVDLKK LRVKELKKIL DDWGEMCKGC AEKSDYIRKI NELMPKYAPK AASARTDL
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers