missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Mucosal addressin cell adhesion molecule 1 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Mucosal addressin cell adhesion molecule 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6331-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16032520

  • 735.00 € / 100µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_062190.1
Mucosal addressin cell adhesion molecule 1 Protein
Research Use Only
Madcam1
Recombinant Protein
E. coli
QSFQVNPPEP EVAVAMGTSL QINCSMSCDK DIARVHWHGL DTNLGNVQTL PGSRVLSVRG MLSDTGTRVC VGSCGSRSFQ HSVKILVYAF PDQLEVTPEF LVPGRDQVVS CTAHNIWPAG PDSLSFALLR GEQSLEGAQA LETEQEEEMQ ETEGTPLFQV TQRWLLPSLG TPALPALYCQ VTMQLPKLVL THRRKIPVLQ SQTSPEPPST TSAKPYILTS SHTTKAVSTG LSSVALPSTP LSSEGPCYPE IHQNPEADWE LLCEASCGSG VTVHWTLAPG DLAAYHKREA GAQAWLSVLP LGPIPEGWFQ CRMDPGGQVT SLYVTGQVIP NPSS
Greater than 90% as determined by SDS-PAGE.
54266
100 μg
Not determined.
Extracellular Domain
Rat
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers