missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Myosin-binding protein C, cardiac-type Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Myosin-binding protein C, cardiac-type, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9297-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16018031

  • 735.00 € / 100µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001099960.1
Myosin-binding protein C, cardiac-type Protein
Research Use Only
Mybpc3
Recombinant Protein
Yeast
PKIHLDCPGS TPDTIVVVAG NKLRLDVPIS GDPAPTVIWQ KTITQGKKAS AGPPPGAPED AGADEEWVFD KKLLCETEGR VRVETTKDRS VFTVEGAEKE DEGVYTVTVK NPVGEDQVNL TVKVIDVPDA PAAPKISNVG EDSCIVQWEP PAYDGGQPVL GYILERKKKK SYRWMRLNFD LLRELSHEAR RMIEGVAYEM RVYAVNAVGM SRPSPASQPF
Greater than 90% as determined by SDS-PAGE.
295929
100 μg
Not determined.
Partial (see sequence information for more details).
Rat
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers