missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat PLA2G2A Protein

A DNA sequence encoding the Rattus norvegicus (Rat) PLA2G2A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7058-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16037260

  • 762.00 € / 100µg
Estimated Shipment: 04-08-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_113786.3
PLA2G2A Protein
100 μg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
29692
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Pla2g2a
Recombinant Protein
Yeast
SLLEFGQMIL FKTGKRADVS YGFYGCHCGV GGRGSPKDAT DWCCVTHDCC YNRLEKRGCG TKFLTYKFSY RGGQISCSTN QDSCRKQLCQ CDKAAAECFA RNKKSYSLKY QFYPNKFCKG KTPSC
Videos
SDS
Documents
Certificates
Special Offers

Special Offers