missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat PRDX2 Protein

A cDNA sequence encoding the PRDX2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13134-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15932609

  • 120.00 € / 5µg
Estimated Shipment: 23-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

PRDX2 Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Rat
His
PRDX2 protein solution (1 mg/ml) containing Phosphate Buffered Saline (pH 7.4), 10% glycerol and 1mM DTT.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMASGNA HIGKPAPDFT GTAVVDGAFK EIKLSDYRGK YVVLFFYPLD FTFVCPTEII AFSDHAEDFR KLGCEVLGVS VDSQFTHLAW INTPRKEGGL GPLNIPLLAD VTKSLSQNYG VLKNDEGIAY RGLFIIDAKG VLRQITVNDL PVGRSVDEAL RLVQAFQYTD EHGEVCPAGW KPGSDTIKPN VDDSKEYFSK HN
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers