missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat PRL R Protein
Specifications
Specifications
| Name | PRL R Protein |
| Quantity | 5 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
| Biological Activity | Activity is determined by the dose-dependant inhibition of Prolactin-stimuled proliferation of Nb2 cells and by high affinity binding of oPLR and other lactogenic hormones. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Rat |
| Species | E. coli |
| Protein Tag | Untagged |
| Sequence | MPSALAFVLL VLNISLLKGQ SPPGKPEIHK CRSPDKETFT CWWNPGTDGG LPTNYSLTYS KEGEKTTYEC PDYKTSGPNS CFFSKQYTSI WKIYIITVNA TNQMGSSSSD PLYVDVTYIV EPEPPRNLTL EVKQLKDKKT YLWVKWSPPT ITDVKTGWFT MEYEIRLKPE EAEEWEIHFT GHQTQFKVFD LYPGQKYLVQ TRCKPDHGYW SRWSQESSVE MPNDFTLKDT TVWIIVAILS AVICLIMVWA VALKGYSMMT CIFPPVPGPK IKGFDTHLLE KGKSEELLSA LGCQDFPPTS DCEDLLVEFL EVDDNEDERL MPSHSKEYPG QGVKPTHLDP DSDSGHGSYD SHSLLSEKCE EPQAYPPTLH IPEITEKPEN PEANIPPTVD PQSTNPNFHV DAPKSSTWPL LPGQHMPRSP YHSVADVCKL AGSPVNTLDS FLDKAEENVL KLSKALETGE EEVAEQKGAK SFPSDKQNTP WPLLQEKSPT VYVKPPDYVE IHKVNKDGVL SLFPKQRENN QTEKPGVPET SKEYAKVSGI TDNNILVLVP DSRAQNTALL EESAKKAPPS FEADQSEKDL ASFTATSSNR RLQLGRLDYL DPTCFMHSFH |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?