missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat PRL R Protein

Product Code. p-7148655
Change view
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15915348 5 μg 5µg
15905348 20 μg 20µg
15995338 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15915348 Supplier enQuireBio™ Supplier No. QP108455ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the PRL R was constructed and used to recombinantly synthesize the protein.

Specifications

Name PRL R Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity Activity is determined by the dose-dependant inhibition of Prolactin-stimuled proliferation of Nb2 cells and by high affinity binding of oPLR and other lactogenic hormones.
Product Type Recombinant Protein
Cross Reactivity Rat
Species E. coli
Protein Tag Untagged
Sequence MPSALAFVLL VLNISLLKGQ SPPGKPEIHK CRSPDKETFT CWWNPGTDGG LPTNYSLTYS KEGEKTTYEC PDYKTSGPNS CFFSKQYTSI WKIYIITVNA TNQMGSSSSD PLYVDVTYIV EPEPPRNLTL EVKQLKDKKT YLWVKWSPPT ITDVKTGWFT MEYEIRLKPE EAEEWEIHFT GHQTQFKVFD LYPGQKYLVQ TRCKPDHGYW SRWSQESSVE MPNDFTLKDT TVWIIVAILS AVICLIMVWA VALKGYSMMT CIFPPVPGPK IKGFDTHLLE KGKSEELLSA LGCQDFPPTS DCEDLLVEFL EVDDNEDERL MPSHSKEYPG QGVKPTHLDP DSDSGHGSYD SHSLLSEKCE EPQAYPPTLH IPEITEKPEN PEANIPPTVD PQSTNPNFHV DAPKSSTWPL LPGQHMPRSP YHSVADVCKL AGSPVNTLDS FLDKAEENVL KLSKALETGE EEVAEQKGAK SFPSDKQNTP WPLLQEKSPT VYVKPPDYVE IHKVNKDGVL SLFPKQRENN QTEKPGVPET SKEYAKVSGI TDNNILVLVP DSRAQNTALL EESAKKAPPS FEADQSEKDL ASFTATSSNR RLQLGRLDYL DPTCFMHSFH
Buffer The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.0045mM NaHCO3.
Purity or Quality Grade Greater than 97.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.