missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat PROS1 / Protein
A DNA sequence encoding the Rattus norvegicus (Rat) PROS1 /, was expressed in the hosts and tags indicated.
580.00 € - 1620.00 €
Specifications
Name | PROS1 / Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | PROS1 |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16054000
|
enQuireBio™
QP6534-MA-20UG |
20 μg |
580.00 €
20µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16044000
|
enQuireBio™
QP6534-MA-100UG |
100 μg |
1620.00 €
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16064000
|
enQuireBio™
QP6534-MA-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
PROS1 / Protein | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
PROS1 | |
Recombinant Protein | |
Mammal | |
ANTLLEETKK GNLERECIEE LCNKEEAREV FENNPETDYF YPKYLGCLGA FRVGAFSAAR QSANAYPDLR SCVNAIPDQC DPMPCNEDGY LSCKDGQGAF TCICKPGWQG DKCQFDINEC KDPSNINGGC SQTCDNTPGS YHCSCKIGFA MLTNKKDCKD VDECSLKPSV CGTAVCKNIP GDFECECPNG YRYDPSSKSC KDVDECSENT CAQLCVNYPG GYSCYCDGKK GFKLAQDQRS CEGIPVCLSL DLDKNYELLY LAEQFAGVVL YLKFRLPDIT RFSAEFDFRT YDSEGIILYA ESLDHSNWLL IALREGKIEV QFKNEFSTQI TTGGNVINNG IWNMVSVEEL DDSVSIKIAK EAVMNINKLG SLFKPTDGFL DTKIYFAGLP RKVESALIKP INPRLDGCIR GWNLMKQGAL GAKEIVEGKQ NKHCFLTVEK GSYYPGSGIA QFSIDYNNVT NAEGWQINVT LNIRPSTGTG VMLALVSGDT VPFALSLVDS GSGTSQDILV FVENSVAAHL EAITLCSEQP SQLKCNINRN GLELWTPVRK DVIYSKDLQR QLAILDKTMK GTVATYLGGV PDISFSATPV NAFYSGCMEV NINGVQLDLD EAISKHNDIR AHSCPSVRKI QKNF | |
Greater than 90% as determined by SDS-PAGE. |