missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Rantes Protein
A cDNA sequence encoding the Rantes was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10858-1mg
Additional Details : Weight : 0.01000kg
Specifications
81780 | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The activity is determined by the ability to chemoattract total human lymphocytes and murine T-cells at a concentration between 1-10ng/ml corresponding to a Specific Activity of 100,000-1,000,000IU/mg. | |
Rat | |
Untagged | |
Lyophilized from a concentrated (1 mg/ml) solution in water containing no additives. |
Rantes Protein | |
Research Use Only | |
Ccl5 | |
Recombinant Protein | |
E. coli | |
SPYGSDTTPC CFAYLSLALP RAHVKEYFYT SSKCSNLAVV FVTRRNRQVC ANPEKKWVQE YINYLEMS | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |