missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat REG3B / Pap1 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) REG3B / Pap1, was expressed in the hosts and tags indicated.
348.00 € - 3590.00 €
Specifications
Accession Number | NP_445741.1 |
---|---|
Gene ID (Entrez) | 24618 |
Name | REG3B / Pap1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16047430
|
enQuireBio™
QP7120-10UG |
10 μg |
348.00 €
10µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16087430
|
enQuireBio™
QP7120-50UG |
50 μg |
493.00 €
50µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16037430
|
enQuireBio™
QP7120-100UG |
100 μg |
820.00 €
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16067430
|
enQuireBio™
QP7120-200UG |
200 μg |
1330.00 €
200µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16077430
|
enQuireBio™
QP7120-500UG |
500 μg |
2260.00 €
500µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16057430
|
enQuireBio™
QP7120-1MG |
1 mg |
3590.00 €
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_445741.1 | |
REG3B / Pap1 Protein | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
24618 | |
Research Use Only | |
Reg3b | |
Recombinant Protein | |
E. coli | |
EDSPKKIPSA RISCPKGSQA YGSYCYALFQ IPQTWFDAEL ACQKRPEGHL VSVLNVAEAS FLASMVKNTG NSYQYTWIGL HDPTLGGEPN GGGWEWSNND IMNYVNWERN PSTALDRGFC GSLSRSSGFL RWRDTTCEVK LPYVCKFTG | |
Greater than 90% as determined by SDS-PAGE. |